ENA245486400 Enamine

4-[4-(chloromethyl)phenoxymethyl]pyridine hydrochloride

  • MDL number MFCD13195979
  • Cas Number : 1210697-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53998
Availability details for
Quantity
  • Availability
  • Add to Cart