ENA245486410 Enamine

4-{5,6-dimethyl-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-2-yl}butanoic acid

  • MDL number MFCD11543519
  • Cas Number : 1038333-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54008
Availability details for
Quantity
  • Availability
  • Add to Cart