ENA245486418 Enamine

1,3-dimethyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD11643091
  • Cas Number : 4050-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54018
Availability details for
Quantity
  • Availability
  • Add to Cart