ENA245486421 Enamine

2-[(oxiran-2-yl)methyl]-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD09731272
  • Cas Number : 140406-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54021
Availability details for
Quantity
  • Availability
  • Add to Cart