ENA245486425 Enamine

(4-methyl-1H-indol-3-yl)methanol

  • MDL number MFCD11212305
  • Cas Number : 1090903-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54031

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54031
Availability details for
Quantity
  • Availability
  • Add to Cart