ENA245486437 Enamine

2-(chloromethyl)-5-(2,4-dichlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD06797383
  • Cas Number : 183368-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54045
Availability details for
Quantity
  • Availability
  • Add to Cart