ENA245486440 Enamine

1-[4-(4-fluorophenyl)oxan-4-yl]methanamine

  • MDL number MFCD08144102
  • Cas Number : 889939-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54048
Availability details for
Quantity
  • Availability
  • Add to Cart