ENA245486446 Enamine

1-methyl-4-[2-(piperidin-4-yl)ethyl]piperazine

  • MDL number MFCD06740643
  • Cas Number : 775288-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54054

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-54054
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart