ENA245486447 Enamine

4-hydroxy-1-(4-methylphenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11133575
  • Cas Number : 1152604-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54056
Availability details for
Quantity
  • Availability
  • Add to Cart