ENA245486456 Enamine

5-(3-chloro-4-fluorophenyl)-1,2,4-triazin-3-amine

  • MDL number MFCD11134053
  • Cas Number : 1094231-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54068
Availability details for
Quantity
  • Availability
  • Add to Cart