ENA245486458 Enamine

bis[(1,5-dimethyl-1H-pyrrol-2-yl)methyl]amine

  • MDL number MFCD13195997
  • Cas Number : 1209276-40-3
  • Molecular Weight : 231.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54070
Availability details for
Quantity
  • Availability
  • Add to Cart