ENA245486459 Enamine

(1-benzylpiperidin-4-yl)methanamine

  • MDL number MFCD04116211
  • Cas Number : 88915-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-90
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54071
Availability details for
Quantity
  • Availability
  • Add to Cart