ENA245486465 Enamine

1-(4-chlorophenyl)-5-methyl-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD13195999
  • Cas Number : 14058-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54079
Availability details for
Quantity
  • Availability
  • Add to Cart