ENA245486468 Enamine

1-(2-phenylcyclopropyl)ethan-1-amine hydrochloride

  • MDL number MFCD13196002
  • Cas Number : 1212189-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54088
Availability details for
Quantity
  • Availability
  • Add to Cart