ENA245486471 Enamine

2-(4-nitrobenzenesulfinyl)acetic acid

  • MDL number MFCD13196003
  • Cas Number : 3406-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54091
Availability details for
Quantity
  • Availability
  • Add to Cart