ENA245486474 Enamine

1-[1-(3-fluorophenyl)cyclohexyl]methanamine

  • MDL number MFCD06213187
  • Cas Number : 771583-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54094
Availability details for
Quantity
  • Availability
  • Add to Cart