ENA245486476 Enamine

1-(4-chlorophenyl)-4-(2-hydroxyethyl)-5-oxo-2,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD13196005
  • Cas Number : 1210492-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54096
Availability details for
Quantity
  • Availability
  • Add to Cart