ENA245486477 Enamine

4-(2-hydroxyethyl)-1-(4-nitrophenyl)-5-oxo-2,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD13196006
  • Cas Number : 1204296-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54097
Availability details for
Quantity
  • Availability
  • Add to Cart