ENA245486479 Enamine

1-(4,6-dimethylpyrimidin-2-yl)-4-(2-hydroxyethyl)-5-oxo-2,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD13196008
  • Cas Number : 1206153-61-8
  • Molecular Weight : 278.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54099
Availability details for
Quantity
  • Availability
  • Add to Cart