ENA245486480 Enamine

ethyl 4-(2-bromophenyl)-2,4-dioxobutanoate

  • MDL number MFCD11131358
  • Cas Number : 65037-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54102
Availability details for
Quantity
  • Availability
  • Add to Cart