ENA245486484 Enamine

3-amino-1-{[3-(trifluoromethyl)phenyl]methyl}urea

  • MDL number MFCD11212630
  • Cas Number : 1094481-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54122
Availability details for
Quantity
  • Availability
  • Add to Cart