ENA245486486 Enamine

N-(piperidin-4-ylmethyl)acetamide

  • MDL number MFCD01595572
  • Cas Number : 71207-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54124

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54124
Availability details for
Quantity
  • Availability
  • Add to Cart