ENA245486492 Enamine

5-(3-fluoro-4-methylphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09809365
  • Cas Number : 1016492-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54131
Availability details for
Quantity
  • Availability
  • Add to Cart