ENA245486520 Enamine

1-methyl-3-(pyridin-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11180222
  • Cas Number : 1152510-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54181

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54181
Availability details for
Quantity
  • Availability
  • Add to Cart