ENA245486521 Enamine

3-chloro-6-ethoxypyridine-2-carboxylic acid

  • MDL number MFCD11147588
  • Cas Number : 1020945-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54183
Availability details for
Quantity
  • Availability
  • Add to Cart