ENA245486523 Enamine

4-({[(furan-2-yl)methyl]amino}(phenyl)methyl)phenol hydrochloride

  • MDL number MFCD13196017
  • Cas Number : 1208889-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54185
Availability details for
Quantity
  • Availability
  • Add to Cart