ENA245486527 Enamine

4-methanesulfonyl-N-methylaniline

  • MDL number MFCD09745015
  • Cas Number : 119871-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54189
Availability details for
Quantity
  • Availability
  • Add to Cart