ENA245486528 Enamine

1-(4-chlorophenyl)-4-hydroxy-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11133588
  • Cas Number : 334832-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54190
Availability details for
Quantity
  • Availability
  • Add to Cart