ENA245486533 Enamine

3-(2-methoxyethoxy)benzonitrile

  • MDL number MFCD09907640
  • Cas Number : 80407-67-6
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54195
Availability details for
Quantity
  • Availability
  • Add to Cart