ENA245486536 Enamine

1-(4-chlorophenyl)-4-methylpentan-1-one

  • MDL number MFCD11105533
  • Cas Number : 236418-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54199
Availability details for
Quantity
  • Availability
  • Add to Cart