ENA245486548 Enamine

[4-(4-chlorophenyl)oxan-4-yl]methanamine hydrochloride

  • MDL number MFCD11043049
  • Cas Number : 1210893-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 315...317
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54225

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54225
Availability details for
Quantity
  • Availability
  • Add to Cart