ENA245486552 Enamine

3-(5-butyl-1,2,4-oxadiazol-3-yl)phenol

  • MDL number MFCD10689502
  • Cas Number : 10185-80-5
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(5-butyl-1,2,4-oxadiazol-3-yl)phenol
Inchi Code 1S/C12H14N2O2/c1-2-3-7-11-13-12(14-16-11)9-5-4-6-10(15)8-9/h4-6,8,15H,2-3,7H2,1H3
Inchi Key BXVMSTSHSMNINW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54230
Availability details for
Quantity
  • Availability
  • Add to Cart