ENA245486554 Enamine

6-bromo-2-[2-(4-ethoxyphenyl)ethenyl]quinoline-4-carboxylic acid

  • Cas Number : 926216-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53753
Availability details for
Quantity
  • Availability
  • Add to Cart