ENA245486558 Enamine

7-fluoro-2-phenylquinoline-4-carboxylic acid

  • MDL number MFCD08444879
  • Cas Number : 132132-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53754
Availability details for
Quantity
  • Availability
  • Add to Cart