ENA245486561 Enamine

2-[(2-benzyl-3-oxo-2,3-dihydro-1H-isoindol-1-yl)formamido]acetic acid

  • MDL number MFCD13196024
  • Cas Number : 1214672-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54242
Availability details for
Quantity
  • Availability
  • Add to Cart