ENA245486563 Enamine

N-(4-aminophenyl)-4-chlorobenzamide

  • MDL number MFCD01027215
  • Cas Number : 23600-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53755

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53755
Availability details for
Quantity
  • Availability
  • Add to Cart