ENA245486569 Enamine

3-amino-1-(1,1-dioxo-1lambda6-thiolan-3-yl)urea

  • MDL number MFCD03038131
  • Cas Number : 507454-65-1
  • Molecular Weight : 193.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54256
Availability details for
Quantity
  • Availability
  • Add to Cart