ENA245486570 Enamine

1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11128116
  • Cas Number : 1152866-69-7
  • Molecular Weight : 257.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C10H6Cl2N2O2/c11-7-1-2-9(8(12)3-7)14-5-6(4-13-14)10(15)16/h1-5H,(H,15,16)
Inchi Key KTVDXAMDPPHMCX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54257

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-54257
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart