ENA245486571 Enamine

3-amino-4-ethoxybenzonitrile

  • MDL number MFCD12189625
  • Cas Number : 72635-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54259
Availability details for
Quantity
  • Availability
  • Add to Cart