ENA245486572 Enamine

1-(2,4-dichlorophenyl)-1H,4H,5H,6H,7H,8H-cyclohepta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09050188
  • Cas Number : 926190-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54260
Availability details for
Quantity
  • Availability
  • Add to Cart