ENA245486574 Enamine

N-(piperidin-4-yl)thiophene-2-carboxamide hydrochloride

  • MDL number MFCD13196029
  • Cas Number : 1211130-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54262
Availability details for
Quantity
  • Availability
  • Add to Cart