ENA245486575 Enamine

1-(1,3-thiazol-2-yl)-1,4-diazepane

  • MDL number MFCD09729966
  • Cas Number : 868065-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54263
Availability details for
Quantity
  • Availability
  • Add to Cart