ENA245486577 Enamine

[(5-phenylthiophen-2-yl)methyl](propan-2-yl)amine

  • MDL number MFCD11629404
  • Cas Number : 1095204-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54265
Availability details for
Quantity
  • Availability
  • Add to Cart