ENA245486579 Enamine

4-(4-methoxyphenyl)oxane-4-carboxylic acid

  • MDL number MFCD04061780
  • Cas Number : 3648-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-164
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54268
Availability details for
Quantity
  • Availability
  • Add to Cart