ENA245486586 Enamine

1-[3-(2,2,2-trifluoroethoxy)propyl]piperazine

  • MDL number MFCD13196031
  • Cas Number : 1208420-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54275
Availability details for
Quantity
  • Availability
  • Add to Cart