ENA245486594 Enamine

5-(aminomethyl)-N,N-dimethylthiophene-2-sulfonamide

  • MDL number MFCD08444347
  • Cas Number : 923246-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54285
Availability details for
Quantity
  • Availability
  • Add to Cart