ENA245486597 Enamine

3-fluoro-4-(2-methoxy-4-methylphenoxy)aniline

  • MDL number MFCD08687841
  • Cas Number : 937598-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54288
Availability details for
Quantity
  • Availability
  • Add to Cart