ENA245486600 Enamine

[4-(2-methoxy-5-methylphenyl)oxan-4-yl]methanamine

  • MDL number MFCD10692698
  • Cas Number : 1152568-32-5
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54291
Availability details for
Quantity
  • Availability
  • Add to Cart