ENA245486604 Enamine

3-chloro-6-(prop-2-en-1-yloxy)pyridine-2-carboxylic acid

  • MDL number MFCD10690050
  • Cas Number : 1092277-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54296
Availability details for
Quantity
  • Availability
  • Add to Cart