ENA245486620 Enamine

1-[4-(4-methoxyphenyl)oxan-4-yl]methanamine

  • MDL number MFCD03447533
  • Cas Number : 440087-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54316
Availability details for
Quantity
  • Availability
  • Add to Cart